Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: AMMECR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025580.1 | NP_001020751.1 | 4, 5 | 865 | ||
NM_001171689.1 | NP_001165160.1 | 7, 8 | 1163 | |||
NM_015365.2 | NP_056180.1 | 5, 6 | 976 | |||
GenBank mRNA | AJ007014.1 | - | 5, 6 | 1063 | ||
AK001703.1 | - | 5, 6 | 499 | |||
AK091430.1 | - | 7, 8 | 1163 | |||
AK314596.1 | - | 5, 6 | 907 | |||
BC060813.1 | - | 4, 5 | 798 | 30344824 | ||
BX648797.1 | - | 4, 5 | 827 | |||
KU178496.1 | - | 4, 5 | 784 | |||
KU178497.1 | - | 5, 6 | 895 |