Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LETMD1 SIRNA Assays ›
Gene Symbol:
LETMD1
Gene Name:
LETM1 domain containing 1
Gene Aliases:
1110019O13Rik, HCCR-1, HCCR-2, HCCR1, HCRR-2
Chromosome Location:
Chr.12: 51048230 - 51068984 on Build GRCh38
UniGene ID:
Hs.655272
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
22911
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LETMD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243689.1
NP_001230618.1
2
216
NM_001300765.1
NP_001287694.1
2
216
NM_015416.4
NP_056231.3
2
216
NR_045017.1
2
91
NR_045018.1
2
216
NR_045019.1
2
216
NR_045020.1
2
216
XM_006719335.1
XP_006719398.1
2
188
XM_006719336.1
XP_006719399.1
2
76
XM_006719339.1
XP_006719402.1
2
188
XM_006719340.1
XP_006719403.1
2
91
XM_011538161.2
XP_011536463.1
2
223
XM_011538162.2
XP_011536464.1
2
224
XM_011538163.2
XP_011536465.1
2
225
XM_011538164.2
XP_011536466.1
2
223
XM_011538165.2
XP_011536467.1
2
223
XM_017019151.1
XP_016874640.1
2
225
XM_017019152.1
XP_016874641.1
2
191
XM_017019153.1
XP_016874642.1
2
225
XM_017019154.1
XP_016874643.1
2
188
XM_017019155.1
XP_016874644.1
2
63
XM_017019156.1
XP_016874645.1
2
226
XM_017019157.1
XP_016874646.1
2
188
XM_017019158.1
XP_016874647.1
2
76
XM_017019159.1
XP_016874648.1
2
226
XM_017019160.1
XP_016874649.1
2
188
XM_017019161.1
XP_016874650.1
2
227
XM_017019162.1
XP_016874651.1
2
197
XM_017019163.1
XP_016874652.1
2
188
XM_017019164.1
XP_016874653.1
2
208
XM_017019165.1
XP_016874654.1
2
188
XM_017019166.1
XP_016874655.1
2
91
XM_017019167.1
XP_016874656.1
2
188
XM_017019168.1
XP_016874657.1
2
188
GenBank mRNA
AF195651.2
-
2
188
AF315598.1
-
2
52
AF329277.1
-
2
215
AK000639.1
-
2
203
AK057866.1
-
2
161
AK127540.1
-
2
177
AK225836.1
-
2
174
AK301898.1
-
2
176
AK301955.1
-
2
91
AK304707.1
-
2
188
AK315119.1
-
2
183
AL050286.1
-
1
196
AY259835.1
-
2
158
AY259836.1
-
2
158
BC064943.1
-
2
162
6009854
BM549678.1
-
188
BX343550.2
-
2
157
BX376670.2
-
2
157
DA682074.1
-
2
91
DC357852.1
-
2
216
Back To Top
More Information
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Process(es)
protein binding
ribosome binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-462rt:80/10.239.167.23:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0