Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161580.1 | NP_001155052.1 | 4 | 606 | ||
NM_001161581.1 | NP_001155053.1 | 4 | 677 | |||
NM_015426.4 | NP_056241.3 | 4 | 606 | |||
XM_011533560.1 | XP_011531862.1 | 4 | 328 | |||
XM_011533561.1 | XP_011531863.1 | 4 | 328 | |||
XM_011533562.2 | XP_011531864.1 | 4 | 328 | |||
XM_011533563.1 | XP_011531865.1 | 4 | 328 | |||
XM_011533564.2 | XP_011531866.1 | 4 | 328 | |||
XM_011533565.2 | XP_011531867.1 | 4 | 328 | |||
XM_017006104.1 | XP_016861593.1 | 4 | 399 | |||
GenBank mRNA | AK075289.1 | - | 4 | 321 | ||
AK315688.1 | - | 4 | 328 | |||
BC007417.2 | - | 3 | 256 | |||
BC110877.1 | - | 4 | 606 | 5588109 | ||
BC119692.2 | - | 4 | 316 | 40116721 | ||
BC119693.2 | - | 4 | 392 |