Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CYFIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037333.2 | NP_001032410.1 | 31 | 6355 | ||
NM_001291721.1 | NP_001278650.1 | 30 | 6277 | |||
NM_001291722.1 | NP_001278651.1 | 32 | 6430 | |||
NM_014376.3 | NP_055191.2 | 31 | 6481 | |||
GenBank mRNA | AF132197.1 | - | 1 | 1492 | ||
AK172724.1 | - | 1 | 2246 | |||
BC026892.2 | - | 1 | 1177 | |||
BC066933.1 | - | 1 | 1678 |