Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC31B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015490.3 | NP_056305.1 | 3 | 251 | ||
GenBank mRNA | AF274863.1 | - | 3 | 247 | ||
AK289975.1 | - | 3 | 233 | |||
AK294551.1 | - | 3 | 367 | |||
BC034946.1 | - | 3 | 219 | |||
BC044569.1 | - | 3 | 212 | |||
BC053949.1 | - | 3 | 212 | |||
BC071580.1 | - | 3 | 272 | |||
BC144313.1 | - | 2 | 192 | 9052833 | ||
BX640914.1 | - | 3 | 297 |