Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015512.4 | NP_056327.4 | 45 | 7382 | ||
XM_017006129.1 | XP_016861618.1 | 47 | 10339 | |||
XM_017006130.1 | XP_016861619.1 | 46 | 10269 | |||
XM_017006131.1 | XP_016861620.1 | 47 | 10339 | |||
XM_017006132.1 | XP_016861621.1 | 47 | 10340 | |||
XM_017006133.1 | XP_016861622.1 | 47 | 10340 | |||
GenBank mRNA | AB037831.3 | - | 45 | 7382 | ||
AB290163.1 | - | 45 | 7382 | |||
AY221994.1 | - | 2 | 230 |