Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HINFP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243259.1 | NP_001230188.1 | 2, 3 | 303 | ||
NM_015517.4 | NP_056332.2 | 3, 4 | 420 | |||
NM_198971.2 | NP_945322.1 | 2, 3 | 303 | |||
XM_011542744.2 | XP_011541046.1 | 2, 3 | 412 | |||
XM_011542745.2 | XP_011541047.1 | 2, 3 | 262 | |||
XM_017017499.1 | XP_016872988.1 | 2, 3 | 360 | |||
XM_017017500.1 | XP_016872989.1 | 2, 3 | 239 | |||
XM_017017501.1 | XP_016872990.1 | 3, 4 | 366 | |||
XM_017017505.1 | XP_016872994.1 | 2, 3 | 254 | |||
XM_017017506.1 | XP_016872995.1 | 2, 3 | 217 | |||
GenBank mRNA | AK056362.1 | - | 2, 3 | 216 | ||
AK300288.1 | - | 2, 3 | 234 | |||
BC001073.2 | - | 2, 3 | 274 | 2820431 | ||
BC001945.1 | - | 2, 3 | 274 | 2820431 | ||
BC012856.1 | - | 2, 3 | 215 | 3871231 | ||
BC017234.2 | - | 2, 3 | 223 | 3537381 | ||
BF726036.1 | - | 2, 3 | 248 | |||
DC326345.1 | - | 3, 4 | 420 |