Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RAI14 SIRNA Assays ›
Gene Symbol:
RAI14
Gene Name:
retinoic acid induced 14
Gene Aliases:
NORPEG, RAI13
Chromosome Location:
Chr.5: 34656328 - 34832612 on Build GRCh38
UniGene ID:
Hs.431400
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
22991
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RAI14
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145520.1
NP_001138992.1
4
452
NM_001145521.1
NP_001138993.1
5
565
NM_001145522.1
NP_001138994.1
4
394
NM_001145523.1
NP_001138995.1
3
326
NM_001145525.1
NP_001138997.1
6
746
NM_015577.2
NP_056392.2
4
394
XM_006714469.2
XP_006714532.1
4
361
XM_011514016.2
XP_011512318.2
3
308
XM_011514017.2
XP_011512319.1
4
485
XM_011514018.1
XP_011512320.1
4
486
XM_011514019.1
XP_011512321.1
5
460
XM_011514020.1
XP_011512322.1
4
419
XM_011514021.1
XP_011512323.1
4
323
XM_011514022.1
XP_011512324.1
4
302
XM_011514023.1
XP_011512325.1
4
344
XM_011514024.1
XP_011512326.1
4
337
XM_011514025.2
XP_011512327.2
3
305
XM_017009334.1
XP_016864823.1
3
503
XM_017009335.1
XP_016864824.1
4
473
XM_017009336.1
XP_016864825.1
4
337
GenBank mRNA
AB037755.1
-
4
452
AF155135.1
-
4
309
AK308306.1
-
5
489
AK314379.1
-
4
283
AY317139.1
-
6
699
AY354204.2
-
3
326
BC052988.1
-
4
310
6164087
DA485361.1
-
4
394
DB036130.1
-
5
531
DN994273.1
-
3
210
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001011947, XM_008760765, XM_006232062, XM_006232063, XM_006232061, and others.
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0