Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161576.2 | NP_001155048.2 | 6, 7 | 1645 | ||
NM_001161577.1 | NP_001155049.1 | 3, 4 | 538 | |||
NM_015589.5 | NP_056404.4 | 7, 8 | 1909 | |||
GenBank mRNA | AB028976.1 | - | 5, 6 | 976 | ||
AK024652.1 | - | 7, 8 | 1602 | |||
AK025923.1 | - | 5, 6 | 800 | |||
BC021060.1 | - | 3, 4 | 455 | |||
BC021726.2 | - | 3, 4 | 493 | |||
BC121173.1 | - | 6, 7 | 1342 | 40122465 | ||
BC121174.1 | - | 6, 7 | 1342 | 40122467 | ||
BI825501.1 | - | 3, 4 | 538 |