Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042549.1 | NP_001036014.1 | 6 | 673 | ||
NM_015471.3 | NP_056286.3 | 5 | 588 | |||
XM_017000968.1 | XP_016856457.1 | 6 | 2084 | |||
XM_017000969.1 | XP_016856458.1 | 6 | 2210 | |||
GenBank mRNA | AF201941.1 | - | 5 | 560 | ||
AF255793.1 | - | 5 | 574 | |||
AK222996.1 | - | 5 | 558 | |||
AK223260.1 | - | 5 | 568 | |||
AK313730.1 | - | 5 | 584 | |||
AL050084.1 | - | 5 | 367 | |||
BC007067.1 | - | 5 | 562 | 3997280 | ||
BC036048.1 | - | 5 | 566 | |||
BU570983.1 | - | 6 | 653 |