Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HINFP SIRNA Assays ›
Gene Symbol:
HINFP
Gene Name:
histone H4 transcription factor
Gene Aliases:
HiNF-P, MIZF, ZNF743
Chromosome Location:
Chr.11: 119121523 - 119135055 on Build GRCh38
UniGene ID:
Hs.504091
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
23060
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HINFP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243259.1
NP_001230188.1
2, 3
318
NM_015517.4
NP_056332.2
3, 4
435
NM_198971.2
NP_945322.1
2, 3
318
XM_011542744.2
XP_011541046.1
2, 3
427
XM_011542745.2
XP_011541047.1
2, 3
277
XM_011542746.1
XP_011541048.1
1, 2
98
XM_017017499.1
XP_016872988.1
2, 3
375
XM_017017500.1
XP_016872989.1
2, 3
254
XM_017017501.1
XP_016872990.1
3, 4
381
XM_017017502.1
XP_016872991.1
5, 6
706
XM_017017503.1
XP_016872992.1
3, 4
471
XM_017017504.1
XP_016872993.1
3, 4
346
XM_017017505.1
XP_016872994.1
2, 3
269
XM_017017506.1
XP_016872995.1
2, 3
232
XM_017017507.1
XP_016872996.1
4, 5
471
XM_017017508.1
XP_016872997.1
3, 4
346
XM_017017509.1
XP_016872998.1
1, 2
98
GenBank mRNA
AK056362.1
-
2, 3
231
AK300288.1
-
2, 3
249
BC001073.2
-
2, 3
289
2820431
BC001945.1
-
2, 3
289
2820431
BC012856.1
-
2, 3
230
3871231
BC017234.2
-
2, 3
238
3537381
BF726036.1
-
2, 3
263
BG821695.1
-
3, 4
352
DC326345.1
-
3, 4
435
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_002729915.This siRNA also targets the following transcripts in Mouse: NM_172162, BY196765, AK049633, CT010239, BC029091, and others.
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
DNA damage checkpoint
G1/S transition of mitotic cell cycle
regulation of transcription involved in G1/S transition of mitotic cell cycle
negative regulation of transcription from RNA polymerase II promoter
in utero embryonic development
DNA repair
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of gene expression
positive regulation of gene expression
negative regulation of gene expression
establishment of protein localization
myoblast differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
protein binding
enzyme binding
histone binding
transcription regulatory region DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline