Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AUTS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127231.2 | NP_001120703.1 | 1 | 998 | ||
NM_001127232.2 | NP_001120704.1 | 1 | 998 | |||
NM_015570.3 | NP_056385.1 | 1 | 998 | |||
XM_011516010.2 | XP_011514312.1 | 1 | 1442 | |||
XM_011516011.2 | XP_011514313.1 | 1 | 1442 | |||
XM_011516012.2 | XP_011514314.1 | 1 | 1442 | |||
XM_011516013.2 | XP_011514315.1 | 1 | 1442 | |||
XM_011516014.2 | XP_011514316.1 | 1 | 1442 | |||
XM_017011951.1 | XP_016867440.1 | 1 | 1442 | |||
GenBank mRNA | AB007902.2 | - | 1 | 559 | ||
AF326917.1 | - | 1 | 584 | |||
BC011643.2 | - | 1 | 306 | |||
BC064693.1 | - | 1 | 1006 | 30345831 | ||
JQ670866.1 | - | 1 | 263 |