Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161576.2 | NP_001155048.2 | 7 | 1745 | ||
NM_001161577.1 | NP_001155049.1 | 4 | 638 | |||
NM_015589.5 | NP_056404.4 | 8 | 2009 | |||
GenBank mRNA | AB028976.1 | - | 6 | 1076 | ||
AK024652.1 | - | 8 | 1702 | |||
AK025923.1 | - | 6 | 900 | |||
BC021060.1 | - | 4 | 555 | |||
BC021726.2 | - | 4 | 593 | |||
BC121173.1 | - | 7 | 1442 | 40122465 | ||
BC121174.1 | - | 7 | 1442 | 40122467 | ||
BI825501.1 | - | 4 | 638 |