Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MXRA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015419.3 | NP_056234.2 | 4, 5 | 874 | ||
XM_005274485.2 | XP_005274542.1 | 4, 5 | 1329 | |||
GenBank mRNA | AF245505.1 | - | 3, 4 | 717 | ||
BC011846.2 | - | 4, 5 | 871 | |||
BC064986.1 | - | 4, 5 | 871 | |||
BC080586.1 | - | 4, 5 | 871 |