Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABI3BP SIRNA Assays ›
Gene Symbol:
ABI3BP
Gene Name:
ABI family member 3 binding protein
Gene Aliases:
NESHBP, TARSH
Chromosome Location:
Chr.3: 100749335 - 100993508 on Build GRCh38
UniGene ID:
Hs.477015
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
23109
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABI3BP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015429.3
NP_056244.2
35
3879
XM_005247281.2
XP_005247338.1
68
6036
XM_005247282.2
XP_005247339.1
67
5994
XM_005247283.2
XP_005247340.1
67
5988
XM_005247284.2
XP_005247341.1
67
5979
XM_005247285.2
XP_005247342.1
67
5976
XM_005247287.2
XP_005247344.1
67
5970
XM_005247288.2
XP_005247345.1
67
5964
XM_005247289.2
XP_005247346.1
67
5961
XM_005247290.2
XP_005247347.1
67
5961
XM_005247291.2
XP_005247348.1
67
5958
XM_005247293.2
XP_005247350.1
66
5928
XM_005247294.2
XP_005247351.1
66
5919
XM_005247295.2
XP_005247352.1
66
5898
XM_005247296.2
XP_005247353.1
66
5895
XM_005247298.2
XP_005247355.1
66
5892
XM_005247299.3
XP_005247356.1
66
5904
XM_005247300.2
XP_005247357.1
65
5862
XM_005247301.2
XP_005247358.1
65
5835
XM_005247302.2
XP_005247359.1
65
5832
XM_005247303.2
XP_005247360.1
64
5775
XM_005247305.2
XP_005247362.1
63
5691
XM_005247306.2
XP_005247363.1
61
5628
XM_005247309.1
XP_005247366.1
66
5669
XM_006713569.2
XP_006713632.1
66
5883
XM_006713570.2
XP_006713633.1
65
5850
XM_006713571.2
XP_006713634.1
65
5820
XM_006713572.2
XP_006713635.1
65
5814
XM_011512639.1
XP_011510941.1
67
5976
XM_011512640.1
XP_011510942.1
67
5970
XM_011512641.1
XP_011510943.1
67
5961
XM_011512643.1
XP_011510945.1
66
5955
XM_011512644.1
XP_011510946.1
67
5952
XM_011512645.1
XP_011510947.1
67
5952
XM_011512646.1
XP_011510948.1
67
5943
XM_011512647.1
XP_011510949.1
66
5901
XM_011512648.1
XP_011510950.1
66
5898
XM_011512649.1
XP_011510951.1
66
5892
XM_011512651.1
XP_011510953.1
65
5835
XM_011512652.1
XP_011510954.1
65
5823
XM_011512653.1
XP_011510955.1
64
5814
XM_011512654.1
XP_011510956.1
65
5805
XM_011512656.1
XP_011510958.1
64
5739
XM_011512657.1
XP_011510959.1
64
5736
XM_011512658.1
XP_011510960.1
63
5724
XM_011512659.1
XP_011510961.1
63
5703
XM_011512660.1
XP_011510962.1
63
5673
XM_017006105.1
XP_016861594.1
67
5979
XM_017006106.1
XP_016861595.1
67
5961
XM_017006107.1
XP_016861596.1
65
5829
XM_017006108.1
XP_016861597.1
63
5739
GenBank mRNA
AB056106.1
-
18
2390
AK093861.1
-
18
2214
AL117664.1
-
10
1689
AL162038.1
-
5
1090
BC030221.1
-
35
3859
5205716
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of cell-substrate adhesion
extracellular matrix organization
collagen binding
heparin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-cnw2s:80/10.239.163.240:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0