Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015496.4 | NP_056311.2 | 13 | 3391 | ||
NM_183009.2 | NP_892121.1 | 13 | 3391 | |||
XM_017013306.1 | XP_016868795.1 | 12 | 3171 | |||
XM_017013307.1 | XP_016868796.1 | 12 | 3214 | |||
XM_017013308.1 | XP_016868797.1 | 11 | 3204 | |||
GenBank mRNA | AB037850.1 | - | 13 | 3315 | ||
AF116724.1 | - | 13 | 3315 | |||
AK000668.1 | - | 1 | 97 | |||
AK022906.1 | - | 5 | 1162 | |||
AL832487.1 | - | 13 | 3332 | |||
BC003701.1 | - | 6 | 1479 | |||
BC069239.1 | - | 6 | 1456 | |||
BC112288.1 | - | 13 | 3403 | 8327756 | ||
BC113380.1 | - | 13 | 3391 | 8322432 | ||
BM462567.1 | - | 2 | 551 | |||
CR627454.1 | - | 2 | 935 |