Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015512.4 | NP_056327.4 | 75 | 12320 | ||
XM_017006129.1 | XP_016861618.1 | 77 | 15277 | |||
XM_017006130.1 | XP_016861619.1 | 76 | 15207 | |||
XM_017006131.1 | XP_016861620.1 | 76 | 15151 | |||
GenBank mRNA | AB037831.3 | - | 75 | 12320 | ||
AB290163.1 | - | 74 | 12515 | |||
AK023766.1 | - | 15 | 2330 | |||
AK074130.2 | - | 22 | 3845 | |||
AL117428.1 | - | 7 | 1039 | |||
AY221994.1 | - | 31 | 5363 |