Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: OSBPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015550.3 | NP_056365.1 | 4 | 719 | ||
NM_145320.2 | NP_663160.1 | 3 | 392 | |||
NM_145321.2 | NP_663161.1 | 3 | 392 | |||
NM_145322.2 | NP_663162.1 | 3 | 392 | |||
NR_104111.1 | 3 | 392 | ||||
NR_104112.1 | 3 | 392 | ||||
XM_005249698.3 | XP_005249755.1 | 4 | 439 | |||
XM_006715681.3 | XP_006715744.1 | 4 | 706 | |||
XM_006715682.3 | XP_006715745.1 | 4 | 725 | |||
XM_006715683.3 | XP_006715746.1 | 4 | 725 | |||
XM_006715684.3 | XP_006715747.1 | 3 | 392 | |||
XM_011515258.2 | XP_011513560.1 | 4 | 516 | |||
XM_011515259.2 | XP_011513561.1 | 5 | 1013 | |||
XM_017011948.1 | XP_016867437.1 | 6 | 876 | |||
XM_017011949.1 | XP_016867438.1 | 5 | 1013 | |||
GenBank mRNA | AB014604.2 | - | 4 | 656 | ||
AF323727.1 | - | 4 | 514 | |||
AF392444.1 | - | 4 | 628 | |||
AF491781.1 | - | 3 | 346 | |||
AF491782.1 | - | 3 | 346 | |||
AF491783.1 | - | 3 | 346 | |||
AF491784.1 | - | 3 | 346 | |||
AF491785.1 | - | 3 | 346 | |||
AF491786.1 | - | 3 | 346 | |||
AL832293.1 | - | 2 | 61 | |||
AY008372.1 | - | 4 | 549 | |||
BC017731.1 | - | 4 | 648 | 3909164 |