Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMADHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015702.2 | NP_056517.1 | 3 | 284 | ||
GenBank mRNA | AF060224.1 | - | 2 | 111 | ||
AF131802.1 | - | 3 | 174 | |||
AF161510.1 | - | 3 | 236 | |||
AK310001.1 | - | 3 | 219 | |||
AK313284.1 | - | 3 | 183 | |||
BC000932.2 | - | 3 | 233 | 3446713 | ||
BC010894.1 | - | 3 | 207 | 4246499 | ||
BC022859.1 | - | 3 | 230 | 3826071 | ||
BC023995.1 | - | 3 | 198 | 4103877 | ||
BG610637.1 | - | 3 | 235 | |||
BU570084.1 | - | 3 | 287 |