Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NAT9 SIRNA Assays ›
Gene Symbol:
NAT9
Gene Name:
N-acetyltransferase 9 (putative)
Gene Aliases:
EBSP, hNATL
Chromosome Location:
Chr.17: 74770529 - 74776425 on Build GRCh38
UniGene ID:
Hs.144058
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
23316
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NAT9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001305077.1
NP_001292006.1
3, 4
350
NM_001305078.1
NP_001292007.1
3, 4
349
NM_001305079.1
NP_001292008.1
3, 4
353
NM_001305080.1
NP_001292009.1
3, 4
353
NM_001305081.1
NP_001292010.1
3, 4
350
NM_001305082.1
NP_001292011.1
3, 4
498
NM_001305083.1
NP_001292012.1
3, 4
350
NM_001305084.1
NP_001292013.1
3, 4
353
NM_001305085.1
NP_001292014.1
3, 4
350
NM_001305086.1
NP_001292015.1
3, 4
350
NM_001305087.1
NP_001292016.1
3, 4
373
NM_001305088.1
NP_001292017.1
2, 3
264
NM_015654.4
NP_056469.2
3, 4
353
NR_130953.1
3, 4
353
NR_130954.1
3, 4
350
XM_005257212.4
XP_005257269.1
2, 3
192
XM_006721813.3
XP_006721876.1
2, 3
640
XM_011524607.2
XP_011522909.1
3, 4
295
XM_017024437.1
XP_016879926.1
3, 4
279
XM_017024438.1
XP_016879927.1
3, 4
284
XM_017024439.1
XP_016879928.1
3, 4
320
XM_017024440.1
XP_016879929.1
2, 3
658
XM_017024441.1
XP_016879930.1
2, 3
652
GenBank mRNA
AK123115.1
-
3, 4
285
AK225602.1
-
3, 4
223
AK308569.1
-
3, 4
256
AK312958.1
-
3, 4
223
AL050269.1
-
3, 4
230
AL537925.3
-
3, 4
430
AY632082.1
-
2, 3
189
BC004195.2
-
3, 4
259
3528894
BC004225.2
-
3, 4
248
3538186
BG702300.1
-
3, 4
286
BI821280.1
-
3, 4
250
BM464121.1
-
3, 4
276
BM909550.1
-
3, 4
252
BP197633.1
-
3, 4
265
BP231776.1
-
3, 4
271
BQ068039.1
-
2, 3
146
BU159218.1
-
3, 4
248
BX333236.2
-
3, 4
263
DA010490.1
-
3, 4
293
DA259097.1
-
3, 4
350
DB171198.1
-
3, 4
267
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
N-terminal protein amino acid acetylation
protein binding
N-acetyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-sq56x:80/10.239.161.141:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline