Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305077.1 | NP_001292006.1 | 7 | 1420 | ||
NM_001305078.1 | NP_001292007.1 | 7 | 1419 | |||
NM_001305079.1 | NP_001292008.1 | 7 | 1346 | |||
NM_001305080.1 | NP_001292009.1 | 7 | 1408 | |||
NM_001305081.1 | NP_001292010.1 | 7 | 1417 | |||
NM_001305082.1 | NP_001292011.1 | 7 | 1568 | |||
NM_001305083.1 | NP_001292012.1 | 7 | 1405 | |||
NM_001305084.1 | NP_001292013.1 | 6 | 1304 | |||
NM_001305085.1 | NP_001292014.1 | 6 | 1301 | |||
NM_001305086.1 | NP_001292015.1 | 6 | 1325 | |||
NM_001305087.1 | NP_001292016.1 | 6 | 1324 | |||
NM_001305088.1 | NP_001292017.1 | 6 | 1257 | |||
NM_015654.4 | NP_056469.2 | 7 | 1423 | |||
NR_130953.1 | 8 | 1427 | ||||
NR_130954.1 | 7 | 1305 | ||||
XM_005257212.4 | XP_005257269.1 | 6 | 1262 | |||
XM_006721813.3 | XP_006721876.1 | 6 | 1710 | |||
XM_017024437.1 | XP_016879926.1 | 7 | 1349 | |||
XM_017024439.1 | XP_016879928.1 | 7 | 1313 | |||
XM_017024441.1 | XP_016879930.1 | 5 | 1603 | |||
GenBank mRNA | AK225602.1 | - | 7 | 1293 | ||
AL050269.1 | - | 7 | 1300 | |||
AY632082.1 | - | 6 | 1259 | |||
BC004195.2 | - | 7 | 1329 | 3528894 | ||
BC004225.2 | - | 7 | 1318 | 3538186 |