Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3YL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159597.2 | NP_001153069.1 | 9 | 1421 | ||
NM_001282682.1 | NP_001269611.1 | 11 | 1526 | |||
NM_001282687.1 | NP_001269616.1 | 12 | 1632 | |||
NM_015677.3 | NP_056492.2 | 10 | 1478 | |||
NR_104223.1 | 12 | 3037 | ||||
NR_104225.1 | 10 | 1669 | ||||
NR_104226.1 | 11 | 2141 | ||||
NR_104227.1 | 11 | 2963 | ||||
GenBank mRNA | AK026507.1 | - | 9 | 1356 | ||
AK026586.1 | - | 10 | 1670 | |||
AK123829.1 | - | 12 | 3219 | |||
AL050373.1 | - | 12 | 1645 | |||
BC008374.1 | - | 10 | 1387 | 3996066 | ||
BC008375.1 | - | 9 | 1348 | 3996120 |