Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADARB1 SIRNA Assays ›
Gene Symbol:
ADARB1
Gene Name:
adenosine deaminase, RNA specific B1
Gene Aliases:
ADAR2, DRABA2, DRADA2, RED1
Chromosome Location:
Chr.21: 45074578 - 45226563 on Build GRCh38
UniGene ID:
Hs.474018
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
23446
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ADARB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001112.3
NP_001103.1
6
1544
NM_001160230.1
NP_001153702.1
6
1544
NM_015833.3
NP_056648.1
6
1544
NM_015834.3
NP_056649.1
6
1544
NR_027672.1
5
609
NR_027673.1
7
1662
NR_027674.1
6
1544
NR_073200.1
6
1544
XM_006723954.2
XP_006724017.1
7
1616
XM_011529425.2
XP_011527727.1
7
2736
XM_011529432.1
XP_011527734.1
5
457
XM_017028242.1
XP_016883731.1
5
3212
XM_017028243.1
XP_016883732.1
8
6701
XM_017028244.1
XP_016883733.1
5
3210
XM_017028245.1
XP_016883734.1
5
3165
XM_017028246.1
XP_016883735.1
8
6701
XM_017028247.1
XP_016883736.1
7
6583
XM_017028248.1
XP_016883737.1
8
1525
XM_017028249.1
XP_016883738.1
9
6744
XM_017028250.1
XP_016883739.1
8
6654
XM_017028251.1
XP_016883740.1
5
1326
XM_017028252.1
XP_016883741.1
5
3212
XM_017028253.1
XP_016883742.1
5
3160
XM_017028254.1
XP_016883743.1
7
6583
XM_017028255.1
XP_016883744.1
7
6581
XM_017028256.1
XP_016883745.1
6
1376
GenBank mRNA
AB194369.1
-
5
587
AB194370.1
-
6
1522
AB194371.1
-
6
1522
AB194372.2
-
6
1522
AB194373.1
-
6
1522
AF001042.1
-
4
1124
AF525422.1
-
7
1640
AF533142.1
-
6
1522
AY135659.1
-
7
1640
BC065545.1
-
6
1381
6014605
U76420.1
-
6
1522
U76421.1
-
6
1522
U76422.1
-
6
1522
U82120.1
-
6
1349
U82121.1
-
4
1109
X99227.1
-
5
1256
X99383.1
-
5
1256
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
adenosine to inosine editing
mRNA processing
negative regulation of cell proliferation
negative regulation of cell migration
negative regulation of protein kinase activity by regulation of protein phosphorylation
positive regulation of viral genome replication
innate immune response
defense response to virus
regulation of cell cycle
double-stranded RNA adenosine deaminase activity
mRNA binding
protein binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-tbml7:80/10.239.166.229:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0