Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128852.1 | NP_001122324.1 | 17 | 2508 | ||
NM_001128853.1 | NP_001122325.1 | 17 | 2511 | |||
NM_001128854.1 | NP_001122326.1 | 17 | 2508 | |||
NM_015908.5 | NP_056992.4 | 17 | 2511 | |||
XM_005250405.2 | XP_005250462.1 | 17 | 2368 | |||
XM_005250406.2 | XP_005250463.1 | 17 | 2365 | |||
XM_005250407.2 | XP_005250464.1 | 17 | 2368 | |||
XM_005250408.2 | XP_005250465.1 | 17 | 2365 | |||
XM_017012290.1 | XP_016867779.1 | 18 | 2693 | |||
XM_017012291.1 | XP_016867780.1 | 18 | 2690 | |||
GenBank mRNA | AF082871.1 | - | 7 | 919 | ||
AF112994.1 | - | 4 | 694 | |||
AF248955.1 | - | 17 | 2474 | |||
AK296131.1 | - | 11 | 1477 | |||
AK302820.1 | - | 17 | 2400 | |||
AL096723.1 | - | 17 | 2462 | |||
BC000082.2 | - | 8 | 1032 | |||
BC069249.1 | - | 15 | 2052 | |||
BC109117.1 | - | 17 | 2402 | 40035609 |