Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000214.2 | NP_000205.1 | 3 | 935 | ||
GenBank mRNA | AF003837.1 | - | 3 | 878 | ||
AF028593.1 | - | 3 | 682 | |||
AK312722.1 | - | 3 | 504 | |||
BC098393.1 | - | 3 | 838 | 30528888 | ||
BC126205.1 | - | 4 | 443 | 8991921 | ||
BC126207.1 | - | 3 | 671 | 8991923 | ||
U61276.1 | - | 3 | 789 | |||
U73936.1 | - | 3 | 832 | |||
U77720.1 | - | 3 | 381 |