Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016013.3 | NP_057097.2 | 2 | 587 | ||
NR_045620.1 | 2 | 587 | ||||
XM_006720555.2 | XP_006720618.1 | 2 | 517 | |||
XM_011521658.1 | XP_011519960.1 | 2 | 451 | |||
XM_011521659.2 | XP_011519961.1 | 2 | 614 | |||
XM_017022293.1 | XP_016877782.1 | 2 | 518 | |||
GenBank mRNA | AF151823.1 | - | 2 | 550 | ||
AK309889.1 | - | 2 | 587 | |||
AK310983.1 | - | 2 | 587 | |||
BC000780.2 | - | 2 | 571 | 3504355 |