Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD17B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025780.2 | NP_001020951.1 | 2 | 849 | ||
NM_016014.3 | NP_057098.2 | 2 | 849 | |||
XM_006717134.3 | XP_006717197.2 | 2 | 300 | |||
XM_006717135.1 | XP_006717198.1 | 2 | 837 | |||
XM_017014787.1 | XP_016870276.1 | 2 | 456 | |||
XM_017014788.1 | XP_016870277.1 | 3 | 1241 | |||
XM_017014789.1 | XP_016870278.1 | 2 | 387 | |||
XM_017014790.1 | XP_016870279.1 | 2 | 837 | |||
XM_017014791.1 | XP_016870280.1 | 2 | 504 | |||
GenBank mRNA | AF151825.1 | - | 2 | 533 | ||
AK293060.1 | - | 2 | 849 | |||
BC038390.1 | - | 2 | 346 | 4748883 | ||
BC044576.1 | - | 2 | 797 | 5285591 |