Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016034.4 | NP_057118.1 | 2, 3 | 247 | ||
NR_051967.1 | 2, 3 | 247 | ||||
NR_051968.1 | 2, 3 | 247 | ||||
XM_006717136.3 | XP_006717199.1 | 3, 4 | 371 | |||
GenBank mRNA | AB055388.1 | - | 2, 3 | 171 | ||
AF151849.1 | - | 2, 3 | 171 | |||
AL555185.3 | - | 2, 3 | 172 | |||
BC008017.1 | - | 2, 3 | 183 | 3534388 | ||
BC013108.2 | - | 2, 3 | 191 | 3924671 | ||
BI260471.1 | - | 2, 3 | 246 | |||
BQ723448.1 | - | 2, 3 | 179 | |||
BX362434.2 | - | 2, 3 | 241 |