Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ASCC1 SIRNA Assays ›
Gene Symbol:
ASCC1
Gene Name:
activating signal cointegrator 1 complex subunit 1
Gene Aliases:
ASC1p50, CGI-18, SMABF2, p50
Chromosome Location:
Chr.10: 72096032 - 72217134 on Build GRCh38
UniGene ID:
Hs.500007
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
23593
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ASCC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198798.2
NP_001185727.1
3
487
NM_001198799.2
NP_001185728.1
3
448
NM_001198800.2
NP_001185729.1
3
448
NR_045564.1
3
527
XM_006717873.3
XP_006717936.1
3
689
XM_011539841.2
XP_011538143.1
3
312
XM_011539842.2
XP_011538144.1
3
689
XM_017016291.1
XP_016871780.1
3
312
XM_017016292.1
XP_016871781.1
3
689
XM_017016293.1
XP_016871782.1
3
312
XM_017016294.1
XP_016871783.1
3
689
XM_017016295.1
XP_016871784.1
3
312
XM_017016296.1
XP_016871785.1
2
213
XM_017016297.1
XP_016871786.1
3
312
XM_017016298.1
XP_016871787.1
3
689
XM_017016299.1
XP_016871788.1
3
312
XM_017016300.1
XP_016871789.1
3
689
XM_017016301.1
XP_016871790.1
3
312
XM_017016302.1
XP_016871791.1
3
689
XM_017016303.1
XP_016871792.1
2
213
GenBank mRNA
AF132952.1
-
4
600
AK023436.1
-
3
295
AK094170.1
-
3
282
AK298302.1
-
3
527
AY013290.1
-
4
600
BC012291.1
-
3
334
3858865
DA297980.1
-
3
425
DA582532.1
-
3
448
DA942671.1
-
3
432
DC361917.1
-
3
529
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
DNA dealkylation involved in DNA repair
transcription, DNA-templated
regulation of transcription, DNA-templated
RNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0