Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002838.4 | NP_002829.3 | 32 | 3818 | ||
NM_080921.3 | NP_563578.2 | 29 | 3335 | |||
XM_006711472.3 | XP_006711535.1 | 31 | 3635 | |||
XM_006711473.3 | XP_006711536.1 | 31 | 3563 | |||
XM_006711474.3 | XP_006711537.1 | 30 | 3419 | |||
GenBank mRNA | AK292131.1 | - | 29 | 3208 | ||
Y00062.1 | - | 30 | 3262 | |||
Y00638.1 | - | 32 | 3685 |