Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015310.3 | NP_056125.3 | 5, 6 | 1940 | ||
NM_206909.2 | NP_996792.1 | 2, 3 | 344 | |||
XM_011544467.2 | XP_011542769.1 | 6, 7 | 2770 | |||
XM_011544468.2 | XP_011542770.1 | 6, 7 | 2775 | |||
XM_011544469.2 | XP_011542771.1 | 6, 7 | 2775 | |||
XM_011544471.2 | XP_011542773.1 | 6, 7 | 2775 | |||
XM_011544473.2 | XP_011542775.1 | 6, 7 | 2775 | |||
XM_017013258.1 | XP_016868747.1 | 5, 6 | 3249 | |||
XM_017013259.1 | XP_016868748.1 | 5, 6 | 3227 | |||
XM_017013260.1 | XP_016868749.1 | 5, 6 | 2000 | |||
XM_017013261.1 | XP_016868750.1 | 4, 5 | 2214 | |||
XM_017013262.1 | XP_016868751.1 | 6, 7 | 2770 | |||
XM_017013263.1 | XP_016868752.1 | 6, 7 | 2775 | |||
XM_017013264.1 | XP_016868753.1 | 5, 6 | 1937 | |||
XM_017013265.1 | XP_016868754.1 | 2, 3 | 750 | |||
GenBank mRNA | AB023159.2 | - | 2, 3 | 325 | ||
AF243495.2 | - | 2, 3 | 347 | |||
AF519767.1 | - | 2, 3 | 295 | |||
AK091317.1 | - | 5, 6 | 1940 | |||
AK315099.1 | - | 2, 3 | 372 | |||
AL832778.1 | - | 4, 5 | 1807 | |||
BC075044.2 | - | 2, 3 | 322 | 30915575 | ||
BC075045.2 | - | 2, 3 | 322 | 30915359 | ||
DB055312.1 | - | 2, 3 | 344 | |||
GU727648.1 | - | 5, 6 | 1840 |