Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129884.2 | NP_001123356.1 | 5 | 796 | ||
NM_001243725.1 | NP_001230654.1 | 4 | 904 | |||
NM_001243726.1 | NP_001230655.1 | 4 | 688 | |||
NM_001243727.2 | NP_001230656.1 | 3 | 522 | |||
NM_001278195.1 | NP_001265124.1 | 5 | 836 | |||
NM_015926.5 | NP_057010.1 | 4 | 744 | |||
NR_024012.2 | 3 | 452 | ||||
NR_103462.1 | 3 | 415 | ||||
NR_103463.1 | 3 | 384 | ||||
XM_006713195.3 | XP_006713258.1 | 4 | 684 | |||
XM_006713197.3 | XP_006713260.1 | 4 | 651 | |||
XM_006713198.2 | XP_006713261.1 | 4 | 719 | |||
XM_011533805.2 | XP_011532107.1 | 4 | 1399 | |||
XM_017006573.1 | XP_016862062.1 | 4 | 904 | |||
GenBank mRNA | AF072733.1 | - | 3 | 642 | ||
AK023997.1 | - | 4 | 687 | |||
AK222938.1 | - | 4 | 656 | |||
AK222949.1 | - | 4 | 656 | |||
AL832608.1 | - | 2 | 3293 | |||
AY358972.1 | - | 3 | 643 | |||
BC001592.1 | - | 4 | 666 | |||
BC008742.2 | - | 4 | 904 | 3346733 | ||
BG326586.1 | - | 3 | 439 | |||
BG772477.1 | - | 3 | 457 | |||
CB142778.1 | - | 3 | 416 | |||
HY149215.1 | - | 3 | 438 |