Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ASCC1 SIRNA Assays ›
Gene Symbol:
ASCC1
Gene Name:
activating signal cointegrator 1 complex subunit 1
Gene Aliases:
ASC1p50, CGI-18, SMABF2, p50
Chromosome Location:
Chr.10: 72096032 - 72217134 on Build GRCh38
UniGene ID:
Hs.500007
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
23687
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ASCC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198798.2
NP_001185727.1
10
1583
NM_001198799.2
NP_001185728.1
13
1825
NM_001198800.2
NP_001185729.1
10
1544
NR_045564.1
10
1623
XM_006717873.3
XP_006717936.1
10
1785
XM_011539841.2
XP_011538143.1
11
1526
XM_011539842.2
XP_011538144.1
11
1903
XM_017016291.1
XP_016871780.1
12
1605
XM_017016292.1
XP_016871781.1
12
1982
XM_017016293.1
XP_016871782.1
11
1487
XM_017016294.1
XP_016871783.1
11
1864
XM_017016295.1
XP_016871784.1
11
1485
XM_017016296.1
XP_016871785.1
9
1268
XM_017016297.1
XP_016871786.1
10
1406
XM_017016298.1
XP_016871787.1
10
1783
XM_017016299.1
XP_016871788.1
9
1288
XM_017016300.1
XP_016871789.1
9
1665
XM_017016301.1
XP_016871790.1
9
1322
XM_017016302.1
XP_016871791.1
9
1699
XM_017016303.1
XP_016871792.1
7
1103
GenBank mRNA
AF132952.1
-
11
1693
AK023436.1
-
10
1391
AK093971.1
-
1
1700
AK094170.1
-
13
1659
AK096409.1
-
9
2212
AK124364.1
-
3
1812
AY013290.1
-
11
1693
BC012291.1
-
10
1430
3858865
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
DNA dealkylation involved in DNA repair
transcription, DNA-templated
regulation of transcription, DNA-templated
RNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0