Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCLAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304445.1 | NP_001291374.1 | 2 | 276 | ||
NM_182551.4 | NP_872357.2 | 2 | 297 | |||
XM_005264244.1 | XP_005264301.1 | 2 | 259 | |||
XM_011532741.2 | XP_011531043.1 | 2 | 248 | |||
XM_011532742.2 | XP_011531044.1 | 1 | 16 | |||
XM_011532745.1 | XP_011531047.1 | 2 | 259 | |||
XM_017003747.1 | XP_016859236.1 | 2 | 259 | |||
GenBank mRNA | AK095284.1 | - | 2 | 262 | ||
AK302562.1 | - | 2 | 229 | |||
AY358702.1 | - | 2 | 266 |