Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APH1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077628.2 | NP_001071096.1 | 7 | 1894 | ||
NM_001243771.1 | NP_001230700.1 | 5 | 2061 | |||
NM_001243772.1 | NP_001230701.1 | 6 | 1684 | |||
NM_016022.3 | NP_057106.2 | 6 | 2232 | |||
NR_045033.1 | 6 | 2096 | ||||
NR_045034.1 | 4 | 1987 | ||||
NR_045035.1 | 5 | 2158 | ||||
XM_017001417.1 | XP_016856906.1 | 8 | 2306 | |||
GenBank mRNA | AF151835.1 | - | 6 | 1934 | ||
AF508787.1 | - | 6 | 1890 | |||
AK075295.1 | - | 6 | 1951 | |||
AK075452.1 | - | 6 | 2040 | |||
AK125685.1 | - | 1 | 1978 | |||
AK298832.1 | - | 6 | 1491 | |||
AK307358.1 | - | 4 | 1794 | |||
AX775955.1 | - | 6 | 1968 | |||
AX775957.1 | - | 7 | 1684 | |||
AY358951.1 | - | 6 | 2036 | |||
BC001230.2 | - | 7 | 1664 | 3457593 | ||
BC008732.2 | - | 6 | 1989 | 3162330 | ||
BC009501.2 | - | 6 | 2023 | 4040230 | ||
BC015568.2 | - | 7 | 1653 | 3356401 | ||
BC017699.1 | - | 6 | 1968 | 4177580 | ||
BC020590.1 | - | 7 | 1684 | 4420942 | ||
BI760743.1 | - | 1 | 292 |