Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACTB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016027.2 | NP_057111.1 | 4 | 568 | ||
GenBank mRNA | AF151841.1 | - | 4 | 511 | ||
AK222699.1 | - | 4 | 521 | |||
AK290201.1 | - | 4 | 530 | |||
BC000878.1 | - | 4 | 557 | 3461825 | ||
BC008505.1 | - | 4 | 557 |