Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016034.4 | NP_057118.1 | 4 | 505 | ||
NR_051967.1 | 5 | 648 | ||||
NR_051968.1 | 5 | 703 | ||||
NR_051969.1 | 4 | 708 | ||||
NR_051970.1 | 4 | 650 | ||||
XM_006717136.3 | XP_006717199.1 | 5 | 629 | |||
GenBank mRNA | AB055388.1 | - | 4 | 429 | ||
AF151849.1 | - | 4 | 429 | |||
AL555185.3 | - | 5 | 573 | |||
BC008017.1 | - | 4 | 441 | 3534388 | ||
BC013108.2 | - | 4 | 449 | 3924671 | ||
BQ723448.1 | - | 5 | 635 | |||
BU169388.1 | - | 4 | 566 | |||
BX362434.2 | - | 4 | 499 |