Gene Symbol: C14orf166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016039.2 | NP_057123.1 | 6 | 651 | ||
GenBank mRNA | AF087877.1 | - | 6 | 682 | ||
AF100755.1 | - | 6 | 682 | |||
AF151857.1 | - | 6 | 682 | |||
AK312004.1 | - | 6 | 615 | |||
AY817175.1 | - | 6 | 521 | |||
BC001722.1 | - | 6 | 603 | 3528725 | ||
DQ525179.1 | - | 6 | 686 |