Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCSH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004483.4 | NP_004474.2 | 5 | 658 | ||
NR_033249.1 | 4 | 594 | ||||
XM_017023136.1 | XP_016878625.1 | 5 | 675 | |||
XM_017023137.1 | XP_016878626.1 | 5 | 618 | |||
GenBank mRNA | BC000790.1 | - | 1 | 582 | 3451361 | |
BC009065.1 | - | 1 | 617 | |||
BC020922.1 | - | 1 | 621 | 4247157 | ||
BF798008.1 | - | 5 | 529 | |||
CN283805.1 | - | 4 | 482 | |||
M69175.1 | - | 5 | 585 |