Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TVP23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316919.1 | NP_001303848.1 | 6 | 1725 | ||
NM_001316920.1 | NP_001303849.1 | 6 | 506 | |||
NM_001316921.1 | NP_001303850.1 | 6 | 795 | |||
NM_001316922.1 | NP_001303851.1 | 6 | 720 | |||
NM_001316923.1 | NP_001303852.1 | 5 | 655 | |||
NM_016078.5 | NP_057162.4 | 6 | 800 | |||
GenBank mRNA | AF151906.1 | - | 6 | 800 | ||
AF223467.1 | - | 6 | 800 | |||
AK128119.1 | - | 3 | 2160 | |||
AK290813.1 | - | 6 | 524 | |||
BC008430.1 | - | 6 | 526 | 4292664 | ||
BQ433266.1 | - | 5 | 421 | |||
BX537929.1 | - | 6 | 516 | |||
BX648167.1 | - | 6 | 1725 | |||
CN390782.1 | - | 5 | 498 | |||
DA067248.1 | - | 6 | 506 | |||
DA773045.1 | - | 5 | 380 |
Gene Symbol: TVP23C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135036.1 | NP_001128508.1 | 6 | 684 | ||
GenBank mRNA | AK310363.1 | - | 6 | 643 |
Gene Symbol: TVP23C-CDRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK303475.1 | - | 5 | 467 |