Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP6R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014931.3 | NP_055746.3 | 7 | 997 | ||
GenBank mRNA | AB029038.2 | - | 7 | 1322 | ||
BC002799.2 | - | 6 | 599 | |||
BC068014.1 | - | 6 | 661 | |||
BC094753.1 | - | 7 | 859 | 4838365 | ||
BX362839.2 | - | 6 | 697 |