Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286587.1 | NP_001273516.1 | 2 | 110 | ||
NM_001286588.1 | NP_001273517.1 | 1 | 167 | |||
NM_001286589.1 | NP_001273518.1 | 1 | 167 | |||
NM_016108.3 | NP_057192.2 | 1 | 167 | |||
XM_011535881.2 | XP_011534183.1 | 1 | 149 | |||
XM_011535882.2 | XP_011534184.1 | 1 | 148 | |||
XM_011535883.2 | XP_011534185.1 | 1 | 148 | |||
XM_011535884.2 | XP_011534186.1 | 1 | 150 | |||
XM_011535885.2 | XP_011534187.1 | 1 | 148 | |||
XM_017010927.1 | XP_016866416.1 | 1 | 166 | |||
XM_017010928.1 | XP_016866417.1 | 1 | 148 | |||
GenBank mRNA | AF151861.1 | - | 2 | 113 | ||
AF153605.1 | - | 1 | 110 | |||
AK001347.1 | - | 1 | 108 | |||
AK222475.1 | - | 1 | 129 | |||
AK298533.1 | - | 1 | 167 | |||
BC025278.1 | - | 1 | 98 | 4859216 | ||
BF725860.1 | - | 1 | 156 | |||
BX538067.1 | - | 1 | 26 | |||
BX640703.1 | - | 1 | 109 |