Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040443.2 | NP_001035533.1 | 3 | 629 | ||
NM_001040444.2 | NP_001035534.1 | 3 | 305 | |||
NR_135322.1 | 2 | 507 | ||||
NR_135323.1 | 4 | 441 | ||||
GenBank mRNA | AB011031.1 | - | 3 | 278 | ||
AF155105.1 | - | 3 | 248 | |||
AK026228.1 | - | 3 | 269 | |||
AK225986.1 | - | 4 | 391 | |||
AK293242.1 | - | 3 | 629 | |||
AK300410.1 | - | 3 | 269 | |||
AL552215.3 | - | 3 | 437 | |||
BC017212.2 | - | 3 | 258 | 4054134 | ||
BX461254.2 | - | 2 | 137 | |||
DA652444.1 | - | 2 | 452 | |||
DB050568.1 | - | 4 | 396 |
Gene Symbol: SETDB2-PHF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320727.1 | NP_001307656.1 | 13 | 2625 | ||
NR_135324.1 | 13 | 2664 | ||||
GenBank mRNA | JF729368.1 | - | 3 | 225 | ||
JF729369.1 | - | 2 | 103 | |||
JF729370.1 | - | 3 | 240 |