Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PAM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016069.9 | NP_057153.8 | 3, 4 | 359 | ||
GenBank mRNA | AF151894.1 | - | 3, 4 | 266 | ||
AF349455.1 | - | 3, 4 | 253 | |||
AK026514.1 | - | 3, 4 | 286 | |||
BC005024.1 | - | 3, 4 | 268 | 3955475 | ||
BG715429.1 | - | 3, 4 | 363 | |||
CR457362.1 | - | 2, 3 | 221 |
Gene Symbol: CORO7-PAM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201479.1 | NP_001188408.1 | 29, 30 | 3433 | ||
GenBank mRNA | AL833954.1 | - | 31, 32 | 3587 |