Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: BOLA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321025.1 | NP_001307954.1 | 2 | 810 | ||
NM_001321026.1 | NP_001307955.1 | 2 | 868 | |||
NM_016074.4 | NP_057158.1 | 2 | 643 | |||
XM_006711348.3 | XP_006711411.1 | 2 | 676 | |||
GenBank mRNA | AF151901.1 | - | 2 | 579 | ||
BC002771.2 | - | 2 | 603 | |||
BC063405.1 | - | 2 | 603 | 5190565 |