Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282168.1 | NP_001269097.1 | 6 | 510 | ||
NM_001282169.1 | NP_001269098.1 | 6 | 671 | |||
NM_016075.3 | NP_057159.2 | 6 | 537 | |||
GenBank mRNA | AF151903.1 | - | 6 | 458 | ||
AK023182.1 | - | 4 | 254 | |||
AK289740.1 | - | 6 | 480 | |||
BC037279.1 | - | 6 | 486 | 4814999 | ||
BC050439.1 | - | 6 | 460 | 5764280 | ||
BX647847.1 | - | 6 | 478 | |||
CB959894.1 | - | 6 | 494 | |||
CR933653.1 | - | 6 | 625 |