Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286045.1 | NP_001272974.1 | 2 | 286 | ||
NM_001286046.1 | NP_001272975.1 | 2 | 286 | |||
NM_016090.3 | NP_057174.1 | 2 | 286 | |||
GenBank mRNA | AB209753.1 | - | 2 | 184 | ||
AF156098.1 | - | 2 | 164 | |||
AK002015.1 | - | 2 | 199 | |||
AK225615.1 | - | 2 | 152 | |||
AK225667.1 | - | 2 | 177 | |||
AK312619.1 | - | 2 | 242 | |||
BC034381.1 | - | 2 | 153 | 5180771 | ||
BC070262.1 | - | 2 | 147 | 4619267 |