Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316935.1 | NP_001303864.1 | 3 | 277 | ||
NM_016126.3 | NP_057210.2 | 3 | 474 | |||
NR_133633.1 | 3 | 474 | ||||
GenBank mRNA | AF100747.1 | - | 3 | 247 | ||
AF416725.1 | - | 2 | 201 | |||
AF417114.1 | - | 2 | 201 | |||
AK225916.1 | - | 3 | 225 | |||
BC005245.1 | - | 3 | 266 | 3838843 | ||
BF967485.1 | - | 3 | 582 | |||
DA133366.1 | - | 3 | 350 | |||
HY179788.1 | - | 3 | 278 | |||
L32869.1 | - | 1 | 49 | |||
X79302.1 | - | 1 | 49 |