Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193286.1 | NP_001180215.1 | 8, 9 | 921 | ||
NM_012237.3 | NP_036369.2 | 9, 10 | 929 | |||
NM_030593.2 | NP_085096.1 | 8, 9 | 921 | |||
NR_034146.1 | 7, 8 | 852 | ||||
XM_006723111.1 | XP_006723174.1 | 10, 11 | 877 | |||
XM_011526654.1 | XP_011524956.1 | 8, 9 | 814 | |||
XM_011526655.1 | XP_011524957.1 | 6, 7 | 629 | |||
XM_017026500.1 | XP_016881989.1 | 8, 9 | 853 | |||
GenBank mRNA | AB009274.1 | - | 8, 9 | 642 | ||
AF083107.2 | - | 9, 10 | 827 | |||
AF095714.1 | - | 8, 9 | 700 | |||
AF131800.1 | - | 7, 8 | 576 | |||
AF160214.1 | - | 8, 9 | 681 | |||
AJ505014.1 | - | 6, 7 | 1398 | |||
AK025876.1 | - | 8, 9 | 707 | |||
AK054642.1 | - | 9, 10 | 760 | |||
AK131390.1 | - | 3, 4 | 1879 | |||
AK225824.1 | - | 8, 9 | 704 | |||
AK290716.1 | - | 7, 8 | 852 | |||
AK314492.1 | - | 9, 10 | 683 | |||
AY030277.1 | - | 8, 9 | 682 | |||
BC003012.1 | - | 8, 9 | 772 | 2820929 | ||
BC003547.1 | - | 8, 9 | 772 | 3536728 | ||
KF032391.1 | - | 6, 7 | 629 |