Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A4GNT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016161.2 | NP_057245.1 | 2 | 578 | ||
XM_017006543.1 | XP_016862032.1 | 2 | 1129 | |||
XM_017006544.1 | XP_016862033.1 | 2 | 699 | |||
GenBank mRNA | AF141315.1 | - | 2 | 556 | ||
AK225668.1 | - | 2 | 578 | |||
BC119639.1 | - | 2 | 473 | 40115688 | ||
BC119640.2 | - | 2 | 473 | 40115689 |